Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5-phenylbenzoxazole 97%, Thermo Scientific™

Catalog No. AC206731000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC206731000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC206731000 Supplier Thermo Scientific Chemicals Supplier No. 206731000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 61931-68-8
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C14H11NO
Quantity 100g
MDL Number MFCD00005772
Synonym 2-methyl-5-phenylbenzoxazole, 2-methyl-5-phenylbenzo d oxazole, benzoxazole, 2-methyl-5-phenyl, 2-methyl-5-phenyl-benzooxazole, 2-methyl-5-phenyl benzoxazole, pubchem8655, acmc-1b9i5, 2-methyl-5-phenylbenzooxazole, 2-methyl-5-phenyl-benzoxazole, benzoxazole,2-methyl-5-phenyl
InChI Key CHZDPBVCBBQQRP-UHFFFAOYSA-N
SMILES CC1=NC2=C(O1)C=CC(=C2)C3=CC=CC=C3
IUPAC Name 2-methyl-5-phenyl-1,3-benzoxazole
Molecular Weight (g/mol) 209.248
PubChem CID 112786
Formula Weight 209.25
Percent Purity ≥96.0%
Chemical Name or Material 2-Methyl-5-phenylbenzoxazole, 97%
Show More Show Less
EINECSNumber 263-330-6
RTECSNumber DM4865170
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.