accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3OH(Z)

CAS RN 61931-81-5

IUPAC Name: (3Z)-hex-3-en-1-yl 2-hydroxypropanoate
Synonyms: cis-3-hexenyl lactate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: NNLLMULULOBXBY-PLNGDYQANA-N
SMILES: CC\C=C/CCOC(=O)C(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results