Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexen-1-yl Lactate 96.0+%, TCI America™
SDP

Catalog No. L010825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L010825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L010825ML Supplier TCI America Supplier No. L010825ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61931-81-5
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00036495
InChI Key NNLLMULULOBXBY-PLNGDYQASA-N
Synonym Lactic Acid cis-3-Hexen-1-yl Ester
PubChem CID 5364231
IUPAC Name [(Z)-hex-3-enyl] 2-hydroxypropanoate
SMILES CCC=CCCOC(=O)C(C)O

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 172.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl Lactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.