accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 619333-95-8

IUPAC Name: tert-butyl N-{[bis(4-methoxyphenyl)phosphoroso]oxy}carbamate
Synonyms: tert-butyl N-{[bis(4-methoxyphenyl)phosphoroso]oxy}carbamate
Molecular Weight (g/mol): 393.38
Molecular Formula: C19H24NO6P
InChi Key: BNMBQDIFMJFRAF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)P(=O)(ONC(=O)OC(C)(C)C)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results