Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl [Bis(4-methoxyphenyl)phosphinyloxy]carbamate 95.0+%, TCI America™
SDP

Catalog No. B28575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28575G 5 g
B28571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28575G Supplier TCI America Supplier No. B28575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 619333-95-8
Molecular Formula C19H24NO6P
Molecular Weight (g/mol) 393.38
MDL Number MFCD08276321
InChI Key BNMBQDIFMJFRAF-UHFFFAOYSA-N
Synonym [Bis(4-methoxyphenyl)phosphinyloxy]carbamic Acid tert-Butyl Ester, O-[Bis(4-methoxyphenyl)phosphinyl]-N-Boc-hydroxylamine, O-[Bis(4-methoxyphenyl)phosphinyl]-N-(tert-butoxycarbonyl)hydroxylamine
PubChem CID 44629804
IUPAC Name tert-butyl N-{[bis(4-methoxyphenyl)phosphoroso]oxy}carbamate
SMILES COC1=CC=C(C=C1)P(=O)(ONC(=O)OC(C)(C)C)C1=CC=C(OC)C=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 393.38
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl [Bis(4-methoxyphenyl)phosphinyloxy]carbamate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.