accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OON

CAS RN 6197-30-4

IUPAC Name: 2-ethylhexyl 2-cyano-3,3-diphenylprop-2-enoate
Synonyms: octocrylene octocrilenum octocrilene octocrileno UV absorber-3 奥克立林 octocrilene (product)
Molecular Weight (g/mol): 361.49
Molecular Formula: C24H27NO2
InChi Key: FMJSMJQBSVNSBF-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C(C#N)=C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)