Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octocrylene, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014913 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4913 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4913 Supplier MilliporeSigma Supelco Supplier No. 023431ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 6197-30-4
Density 1.051 g/mL (at 25°C)
Flash Point 234°C - closed cup
Linear Formula (C6H5)2 C=C(CN)CO2 CH2 CH(C2H5)(CH2)3 CH3
MDL Number MFCD00059260
Melting Point -10°C
Boiling Point 218°C (1.5 mmHg)
Synonym 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate
Type Analytical Standard
Quantity 1 mL
Formula Weight 361.48
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.