accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHNO

CAS RN 61977-29-5

IUPAC Name: 6-chloro-2,3-dihydro-1,2-benzoxazol-3-one
Synonyms: 6-chloro-2H-1,2-benzoxazol-3-one
Molecular Weight (g/mol): 169.56
Molecular Formula: C7H4ClNO2
InChi Key: SJAPSPJRTQCDNO-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(=O)NOC2=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results