accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNHOOHHO

CAS RN 62-13-5

IUPAC Name: hydrogen 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethan-1-one chloride
Synonyms: adrenone hydrochloride
Molecular Weight (g/mol): 217.65
Molecular Formula: C9H12ClNO3
InChi Key: CSRRBDMYOUQTCO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCC(=O)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)