Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3',4'-Dihydroxy-2-(methylamino)acetophenone hydrochloride

Catalog No. 0227010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-270-10G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-270-10G Supplier Sigma Aldrich Supplier No. 0227010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The adsorption data of propyl benzoate was obtained by frontal analysis on a symmetry-C18 column by a mixture of methanol and water as the mobile phase.Propyl benzoate was used to extract desipramine, a model compound used in the electrochemical analysis by a combined hollow fiber-based liquid three-phase microextraction and voltammetric method.

Specifications

CAS 62-13-5
Melting Point 244°C to 249°C (dec.)
Molecular Formula C9H11NO3 -+ HCl
Linear Formula C9H11NO3 -+ HCl
MDL Number MFCD00035075
Quantity 50 g
Synonym Adrenalone hydrochloride
Molecular Weight (g/mol) 217.65
Percent Purity ≥98.0% (calc. based on dry substance, AT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.