accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N

CAS RN 620-22-4

IUPAC Name: 3-methylbenzonitrile
Synonyms: benzonitrile, 3-methyl-
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: BOHCMQZJWOGWTA-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C#N
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)