Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Tolunitrile, 94%, Thermo Scientific™

Catalog No. AAL1497730
Change view
Click to view available options
Quantity:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1497730 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1497730 Supplier Thermo Scientific Chemicals Supplier No. L1497730

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 620-22-4
Boiling Point 210°C
Flash Point 86°C (186°F)
Odor Almond-like
Molecular Formula C8H7N
Quantity 250g
MDL Number MFCD00001808
Beilstein 507391
Synonym m-tolunitrile, 3-cyanotoluene, benzonitrile, 3-methyl, m-cyanotoluene, 3-tolunitrile, m-toluonitrile, m-tolylnitrile, m-toluenenitrile, m-methylbenzonitrile, m-tolynitrile
InChI Key BOHCMQZJWOGWTA-UHFFFAOYSA-N
SMILES CC1=CC=CC(=C1)C#N
IUPAC Name 3-methylbenzonitrile
Molecular Weight (g/mol) 117.151
PubChem CID 12104
Formula Weight 117.15
Percent Purity 94%
Chemical Name or Material m-Tolunitrile
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECSNumber XV0525000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.