accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6203-18-5

IUPAC Name: (2E)-3-[4-(dimethylamino)phenyl]prop-2-enal
Synonyms: P-dimethylaminocinnamaldehyde
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: RUKJCCIJLIMGEP-ONEGZZNKSA-N
SMILES: CN(C)C1=CC=C(\C=C\C=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (4)
Quantity 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (3)