Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)Cinnamaldehyde, Sigma-Aldrich

Catalog No. 01258837 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-837 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-837 Supplier MilliporeSigma Supelco Supplier No. D450625G
Add to Cart
Add to Cart
  • 4-(Dimethylamino)cinnamaldehyde (DMAC) has been used in the serine protease inhibition assay.
  • It has been used for the detection of (epi)-flavan-3-ols and proanthocyanidin polymers in Brassica napus seeds.
  • It has been used for the determination of phenolic compounds in leaf samples.
  • It has been used for the staining of flavan-3-ols in plant samples.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6203-18-5
Molecular Weight (g/mol) 175.23 g/mol

Specifications

Quantity 25 g
Percent Purity ≥98%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.