accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOO

CAS RN 6207-89-2

IUPAC Name: 1,3-dioxo-1H,2H,3H-benzo[e]isoindol-2-olate
Synonyms: 1,3-dioxobenzo[e]isoindol-2-olate
Molecular Weight (g/mol): 212.19
Molecular Formula: C12H6NO3
InChi Key: WIBMBVLFXLIGEE-UHFFFAOYSA-N
SMILES: [O-]N1C(=O)C2=CC=C3C=CC=CC3=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results