Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxy-1,8-naphthalimide sodium salt, 99%, Thermo Scientific™

Catalog No. AAA1602706
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1602706 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1602706 Supplier Thermo Scientific Chemicals Supplier No. A1602706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6207-89-2
Molecular Formula C12H6NNaO3
Quantity 5g
MDL Number MFCD00006923
Beilstein 4282212
Synonym n-hydroxy-1,8-naphthalimide sodium salt, unii-m4q6d69sag, m4q6d69sag, n,n-1,8-naphthalyl hydroxylamine sodium salt, sodium naphthalhydroxamic acid, n-hydroxynaphthalimide, sodium, naphthalimide, n-hydroxy-, sodium, n-hydroxy-1,8-naphthalimide, sodium, n-sodiooxy-1,8-naphthalenedicarbimide, sodium 1,3-dioxo-1h-benzo de isoquinolin-2 3h-olate
InChI Key VBWKQEZAOAAGNK-UHFFFAOYSA-N
SMILES C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)[O-].[Na+]
IUPAC Name sodium;2-oxidobenzo[de]isoquinoline-1,3-dione
Molecular Weight (g/mol) 235.174
PubChem CID 23689369
Formula Weight 235.18
Percent Purity 99%
Chemical Name or Material N-Hydroxy-1,8-naphthalimide sodium salt
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.