accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHHCl

CAS RN 62075-23-4

IUPAC Name: 4-aminoadamantan-1-ol hydrochloride
Synonyms: 4-aminoadamantan-1-ol hydrochloride
Molecular Weight (g/mol): 203.71
Molecular Formula: C10H18ClNO
InChi Key: KWEPNQFVPHYHHO-UHFFFAOYNA-N
SMILES: Cl.NC1C2CC3CC1CC(O)(C3)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results