Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Amino-1-adamantanol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A25945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25945G Supplier TCI America Supplier No. A25945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62075-23-4
Molecular Formula C10H18ClNO
Molecular Weight (g/mol) 203.71
MDL Number MFCD11099880
InChI Key KWEPNQFVPHYHHO-VSLJLWNASA-N
Synonym trans-1-Amino-4-hydroxyadamantane Hydrochloride
PubChem CID 45489843
IUPAC Name (3S)-4-aminoadamantan-1-ol;hydrochloride
SMILES C1C2CC3CC(C2)(CC1C3N)O.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 203.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Amino-1-adamantanol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.