accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOO

CAS RN 621-36-3

IUPAC Name: 2-(3-methylphenyl)acetic acid
Synonyms: 3-methylphenylacetic acid M-tolylacetic acid
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: GJMPSRSMBJLKKB-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CC(O)=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (1)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (1)
  • (7)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)