Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Tolylacetic acid, 97%

Catalog No. AC139112500
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139112500 250 g Glass bottle
AC139110050 5 g Glass bottle
AC139110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC139112500 Supplier Thermo Scientific Chemicals Supplier No. 139112500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 621-36-3 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-36-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00004340
InChI Key GJMPSRSMBJLKKB-UHFFFAOYSA-N
Synonym 3-methylphenylacetic acid, m-tolylacetic acid, 2-m-tolyl acetic acid, 2-3-methylphenyl acetic acid, 3-tolylacetic acid, m-methylphenylacetic acid, 3-methylphenyl acetic acid, m-tolylaceticacid, benzeneacetic acid, 3-methyl, methyll 3-methylphenylacetate
PubChem CID 12121
IUPAC Name 2-(3-methylphenyl)acetic acid
SMILES CC1=CC=CC(CC(O)=O)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61.0°C to 66.0°C
Color Colorless to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2CO2H
Quantity 250 g
Beilstein 09,528
Formula Weight 150.18
Percent Purity 97%
Physical Form Shiny Flakes and Chunks
Chemical Name or Material m-Tolylacetic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-683-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.