accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-63-6

IUPAC Name: 2,2-diethoxyethan-1-ol
Synonyms: ethanol, 2,2-diethoxy-
Molecular Weight (g/mol): 134.18
Molecular Formula: C6H14O3
InChi Key: IKKUKDZKIIIKJK-UHFFFAOYSA-N
SMILES: CCOC(CO)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)