accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-91-0

IUPAC Name: 1,4-bis(benzyloxy)benzene
Synonyms: benzyl hydroquinone
Molecular Weight (g/mol): 290.36
Molecular Formula: C20H18O2
InChi Key: DYULYMCXVSRUPB-UHFFFAOYSA-N
SMILES: C(OC1=CC=C(OCC2=CC=CC=C2)C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results