Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibenzyloxybenzene 98.0+%, TCI America™
SDP

Catalog No. D01595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01595G 5 g
D015925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01595G Supplier TCI America Supplier No. D01595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 621-91-0
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.36
MDL Number MFCD00016875
InChI Key DYULYMCXVSRUPB-UHFFFAOYSA-N
Synonym 1,4-dibenzyloxybenzene, 1,4-bis benzyloxy benzene, hydroquinone dibenzyl ether, benzene, 1,4-bis phenylmethoxy, 1,4-bis phenylmethoxy benzene, p-dibenzyloxybenzene, hydrochinon-dibenzylather, 1,4-di benzyloxy benzene, acmc-209n0i, ksc493o6b
PubChem CID 69315
IUPAC Name 1,4-bis(benzyloxy)benzene
SMILES C(OC1=CC=C(OCC2=CC=CC=C2)C=C1)C1=CC=CC=C1

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 5 g
Formula Weight 290.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibenzyloxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.