accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62127-47-3

IUPAC Name: (3R)-2,2,3-tribromobutane
Synonyms: (3R)-2,2,3-tribromobutane
Molecular Weight (g/mol): 294.81
Molecular Formula: C4H7Br3
InChi Key: JDWNTZIECWWBAO-GSVOUGTGSA-N
SMILES: C[C@@H](Br)C(C)(Br)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results