Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3-Tribromobutane 96.0+%, TCI America™
SDP

Catalog No. T17695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17695G Supplier TCI America Supplier No. T17695G

Chemical Identifiers

CAS 62127-47-3
Molecular Formula C4H7Br3
Molecular Weight (g/mol) 294.812
MDL Number MFCD00039172
InChI Key JDWNTZIECWWBAO-UHFFFAOYSA-N
PubChem CID 112468
IUPAC Name 2,2,3-tribromobutane
SMILES CC(C(C)(Br)Br)Br

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 294.81
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3-Tribromobutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.