accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COONH(S)IOHO

CAS RN 62129-44-6

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-iodophenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(4-iodophenyl)propanoic acid
Molecular Weight (g/mol): 391.21
Molecular Formula: C14H18INO4
InChi Key: JZLZDBGQWRBTHN-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(I)C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)