Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl Diphenylphosphinite

Catalog No. 1494975G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-949-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 14-949-75G Supplier Sigma Aldrich Supplier No. 1494975G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ased as:Catalyst for hydroformylative desymmetrization of bisalkenyl carbinols and bishomoallylic carbinolsLigand in rhodium-catalyzed hydroformylation of bishomoallylic alcoholsCalatyst in preparation of +a-lactones via branched-regioselective hydroformylation reactionsLigand for nickel-catalyzed hydrocyanation reactionsCatalyst in annulation reactions

Specifications

CAS 62129-44-6
Melting Point ∼150°C
Molecular Formula C14H18INO4
Linear Formula (CH3)3COCONHCH(COOH)CH2C6H4I
Refractive Index n20/D 1.604
MDL Number MFCD00037119
Quantity 5 g
Synonym Diphenylmethoxyphosphine; Methoxydiphenylphosphine; NSC 171167
Molecular Weight (g/mol) 391.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.