CAS RN 6214-18-2
IUPAC Name:
2-methylpropyl benzenesulfonate
Synonyms:
2-methylpropyl benzenesulfonate
Molecular Weight (g/mol):
214.28
Molecular Formula:
C10H14O3S
InChi Key:
NFRRFEFZKGNYKS-UHFFFAOYSA-N
SMILES:
CC(C)COS(=O)(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Benzenesulfonic acid isopropyl ester, 95%
Benzenesulfonic acid isopropyl ester, 95%
CAS: 6214-18-2 Molecular Formula: C9H12O3S Molecular Weight (g/mol): 200.252 MDL Number: MFCD04038771 InChI Key: YQZZXXKFKTWDPY-UHFFFAOYSA-N PubChem CID: 12441430 IUPAC Name: propan-2-yl benzenesulfonate SMILES: CC(C)OS(=O)(=O)C1=CC=CC=C1