accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62140-67-4

IUPAC Name: methyl 5-(ethanesulfonyl)-2-methoxybenzoate
Synonyms: methyl 5-(ethanesulfonyl)-2-methoxybenzoate
Molecular Weight (g/mol): 258.29
Molecular Formula: C11H14O5S
InChi Key: UUARTHQJSZTOEM-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)C1=CC=C(OC)C(=C1)C(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results