accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ClNOO

CAS RN 6216-91-7

IUPAC Name: 2-chloro-4-nitro-N,N-dipropylaniline
Synonyms: 2-chloro-4-nitro-N,N-dipropylaniline
Molecular Weight (g/mol): 256.73
Molecular Formula: C12H17ClN2O2
InChi Key: OYJJPFCGGNTTTO-UHFFFAOYSA-N
SMILES: CCCN(CCC)C1=CC=C(C=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results