accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOO(Z)(Z)(Z)(Z)(Z)(Z)

CAS RN 6217-54-5

IUPAC Name: (4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
Synonyms: docosahexaenoic acid 二十二碳六烯酸 doconexento doconexent doconexentum all-Z-docosahexaenoate DHA doxonexent all-Z-docosahexaenoic acid all-cis-DHA
Molecular Weight (g/mol): 328.5
Molecular Formula: C22H32O2
InChi Key: MBMBGCFOFBJSGT-KUBAVDMBSA-N
SMILES: CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)