Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4,7,10,13,16,19-Docosahexaenoic Acid, >99%, MP Biomedicals™

Catalog No. ICN15909780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15909780 100 mg
ICN15909725 25 mg
ICN15909710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15909780 Supplier MP Biomedicals, Inc Supplier No. 0215909780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.496
MDL Number MFCD00065722
InChI Key MBMBGCFOFBJSGT-OBOJEMQYSA-N
PubChem CID 57417355
IUPAC Name (4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O

Specifications

Color Colorless
Assay Percent Range ≥98%
Quantity 100 mg
Synonym cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
Formula Weight 328.5
Percent Purity ≥98%
Grade Not Available
Physical Form Liquid
Chemical Name or Material cis-4,7,10,13,16,19-Docosahexaenoic Acid
Recommended Storage Store at - 0°C, protect from light, store under nitrogen
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.