accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-15-1

IUPAC Name: N,N'-diphenylmethanimidamide
Synonyms: N,N'-diphenylmethanimidamide
Molecular Weight (g/mol): 196.25
Molecular Formula: C13H12N2
InChi Key: ZQUVDXMUKIVNOW-UHFFFAOYSA-N
SMILES: N(C=NC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)