Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diphenylformamidine 98.0+%, TCI America™
SDP

Catalog No. D122125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D122125G 25 g
D1221250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D122125G Supplier TCI America Supplier No. D122125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 622-15-1
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.25
MDL Number MFCD00003026
InChI Key ZQUVDXMUKIVNOW-UHFFFAOYSA-N
Synonym n,n'-diphenylformamidine, diphenylformamidine, methanimidamide, n,n'-diphenyl, formamidine, n,n'-diphenyl, n,n'-diphenylformamidene, n,n'-diphenylformimidamide, n,n'-diphenymethanimidamide, n,n-diphenylformamidine, formamidine, n,n'-diphenyl-6ci,7ci,8ci, 1e-2-phenyl-2-azavinyl phenylamine
PubChem CID 94888
IUPAC Name N,N'-diphenylmethanimidamide
SMILES N(C=NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 138°C
Quantity 25 g
Formula Weight 196.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diphenylformamidine
EINECSNumber (3)-0375
RTECSNumber PB7705000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.