accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-31-1

IUPAC Name: (E)-N-(phenylmethylidene)hydroxylamine
Synonyms: benzaldoxime,syn
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: VTWKXBJHBHYJBI-SOFGYWHQSA-N
SMILES: O\N=C\C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)