Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (E)-Benzaldehyde oxime

Catalog No. 24567450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-567-450G 50 g
24-567-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-567-450G Supplier Sigma Aldrich Supplier No. 24567450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(E)-Benzaldehyde oxime

Specifications

CAS 622-31-1
Melting Point 34°C to 36°C (lit.)
Boiling Point 104°C (6 mmHg)
Molecular Formula C7H7NO
Linear Formula C6H5CH=NOH
Refractive Index n20/D 1.591 (literature)
Quantity 50 g
MDL Number MFCD00002119
Molecular Weight (g/mol) 121.14
Percent Purity 97% (mixture of cis and trans)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.