accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-46-8

IUPAC Name: phenyl carbamate
Synonyms: O-phenyl carbamate
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: BSCCSDNZEIHXOK-UHFFFAOYSA-N
SMILES: NC(=O)OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)