accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNSOONH2HNO(E/Z)

CAS RN 622387-85-3

IUPAC Name: 3-[(1-methyl-1H-indol-3-yl)methylidene]-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide
Synonyms: 3-[(1-methylindol-3-yl)methylidene]-2-oxo-1H-indole-5-sulfonamide
Molecular Weight (g/mol): 353.4
Molecular Formula: C18H15N3O3S
InChi Key: MLKHXLFEYOOYEY-UHFFFAOYSA-N
SMILES: CN1C=C(C=C2C(=O)NC3=C2C=C(C=C3)S(N)(=O)=O)C2=CC=CC=C12
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results