CAS RN 62254-74-4
IUPAC Name:
5-methyl-1,2-oxazole-3-carbaldehyde
Synonyms:
5-methyl-1,2-oxazole-3-carbaldehyde
Molecular Weight (g/mol):
111.1
Molecular Formula:
C5H5NO2
InChi Key:
OCOCVXUEFWNRJU-UHFFFAOYSA-N
SMILES:
CC1=CC(C=O)=NO1
1
–
1
of
1
results
1
Sigma Aldrich 5-Methylisoxazole-3-carboxaldehyde
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.