accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 623-08-5

IUPAC Name: N,4-dimethylaniline
Synonyms: N-methyl P-toluidine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: QCIFLGSATTWUQJ-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results