Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-p-toluidine 97.0+%, TCI America™
SDP

Catalog No. M131525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M131525ML 25 mL
M1315250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M131525ML Supplier TCI America Supplier No. M131525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 623-08-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00025628
InChI Key QCIFLGSATTWUQJ-UHFFFAOYSA-N
Synonym N,4-Dimethylaniline, 4-(Methylamino)toluene
PubChem CID 12165
IUPAC Name N,4-dimethylaniline
SMILES CNC1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 212°C
Quantity 25 mL
UN Number 2810
Formula Weight 121.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-p-toluidine
EINECSNumber (3)-0129&(3)-0187
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.