accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 623-39-2

IUPAC Name: 3-methoxypropane-1,2-diol
Synonyms: glycerin methyl ether
Molecular Weight (g/mol): 106.12
Molecular Formula: C4H10O3
InChi Key: PSJBSUHYCGQTHZ-UHFFFAOYNA-N
SMILES: COCC(O)CO
Boiling Point 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)