Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-1,2-propanediol, 97%, Thermo Scientific™

Catalog No. AAL0801103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0801103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0801103 Supplier Thermo Scientific Chemicals Supplier No. L0801103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-39-2
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00004719
InChI Key PSJBSUHYCGQTHZ-UHFFFAOYNA-N
Synonym 3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
PubChem CID 92822
IUPAC Name 3-methoxypropane-1,2-diol
SMILES COCC(O)CO

Specifications

Density 1.117
Boiling Point 218°C to 220°C
Flash Point >110°C (230°F)
Refractive Index 1.444
Quantity 1 g
Beilstein 1733242
Sensitivity Hygroscopic
Formula Weight 106.12
Percent Purity 97%
Chemical Name or Material 3-Methoxy-1,2-propanediol
EINECSNumber 210-791-6
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.