accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOHNCH3O

CAS RN 623-59-6

IUPAC Name: 3-acetyl-1-methylurea
Synonyms: 3-acetyl-1-methylurea
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: XRVHSOXXNQTWAW-UHFFFAOYSA-N
SMILES: CNC(=O)NC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results