Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-3-methylurea 98.0+%, TCI America™
SDP

Catalog No. A010125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A010125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A010125G Supplier TCI America Supplier No. A010125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 623-59-6
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00026167
InChI Key XRVHSOXXNQTWAW-UHFFFAOYSA-N
PubChem CID 69337
IUPAC Name N-(methylcarbamoyl)acetamide
SMILES CC(=O)NC(=O)NC

Specifications

Melting Point 180°C
Color White
Quantity 25 g
Formula Weight 116.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetyl-3-methylurea
RTECSNumber YR6850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.