accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 623-65-4

IUPAC Name: hexadecanoyl hexadecanoate
Synonyms: hexadecanoic acid, anhydride
Molecular Weight (g/mol): 494.85
Molecular Formula: C32H62O3
InChi Key: QWZBEFCPZJWDKC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)