Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitic Anhydride 96.0+%, TCI America™
SDP

Catalog No. P0008500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0008500G 500 g
P000825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0008500G Supplier TCI America Supplier No. P0008500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 623-65-4
Molecular Formula C32H62O3
Molecular Weight (g/mol) 494.845
MDL Number MFCD00008992
InChI Key QWZBEFCPZJWDKC-UHFFFAOYSA-N
Synonym palmitic anhydride, hexadecanoic anhydride, palmitic acid anhydride, palmiticanhydride, hexadecanoic acid, anhydride, unii-33jhb8gs96, hexadecanoic acid, 1,1'-anhydride, bispalmitic anhydride, n-hexadecanoic anhydride, hexadecanoic anhydride #
PubChem CID 69339
IUPAC Name hexadecanoyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 64°C
Color White
Quantity 500 g
UN Number 3261
Formula Weight 494.85
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Palmitic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.