accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3C(E)(E)

CAS RN 623-68-7

IUPAC Name: (2E)-but-2-enoyl (2E)-but-2-enoate
Synonyms: 2-butenoic acid, anhydride
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: VJDDQSBNUHLBTD-GGWOSOGESA-N
SMILES: C\C=C\C(=O)OC(=O)\C=C\C
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results