Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonic Anhydride, MP Biomedicals

Catalog No. ICN211133 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211133 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211133 Supplier MP Biomedicals, Inc Supplier No. 0521113310
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-68-7
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
InChI Key VJDDQSBNUHLBTD-GGWOSOGESA-N
Synonym crotonic anhydride, 2-butenoic anhydride, 2-butenoic acid, anhydride, crotonic acid anhydride, but-2-enoic anhydride, unii-n521pmq949, anhydrid kyseliny krotonove czech, crotonicanhydride, e-but-2-enoyl e-but-2-enoate, anhydrid kyseliny krotonove
PubChem CID 6049834
IUPAC Name [(E)-but-2-enoyl] (E)-but-2-enoate
SMILES CC=CC(=O)OC(=O)C=CC

Specifications

Quantity 10 g
Formula Weight 154.17
Chemical Name or Material Crotonic Anhydride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.