accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOOCH3

CAS RN 623-81-4

IUPAC Name: diethyl sulfite
Synonyms: diethyl sulfite
Molecular Weight (g/mol): 138.18
Molecular Formula: C4H10O3S
InChi Key: NVJBFARDFTXOTO-UHFFFAOYSA-N
SMILES: CCOS(=O)OCC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)